Order Entry
Belgium
Orders LinkContactUsLinkComponent
Pipette tips

Pipette tips

Type L, 300 µl, -, Transparent, Eppendorf Reference, Research/Research Plus, mLine Mechanical, Proline Mechanical, eLine Electronic, 96/box, 1920/case
Catalog # SARS70.765.100
Marketsource
Supplier:   SARSTEDT
Pipette tips
Catalog # SARS70.765.100
Supplier:  SARSTEDT
Marketsource
Pipette tips

Type L, 300 µl, -, Transparent, Eppendorf Reference, Research/Research Plus, mLine Mechanical, Proline Mechanical, eLine Electronic, 96/box, 1920/case

Price
Inquire For Price
Log in for availability
1 * 1.920 ST
Product Details & Documents

These high quality tips are suitable for the most common pipettes.

  • Sterile tips are DNase/RNase- and ATP-free, and non pyrogenic
  • Available loose in bags, packed in racks or in Tip StackPack reload systems

Tip StackPack: Tips are preloaded in reusable, stacked 96-tip trays. The trays are designed to safely cover each individual 96-tip layer, shielding the preloaded tips in the stack like a protective screen.

Specifications
  • Packed:
    96/box, 1920/case
  • Sterile:
    -
  • Volume:
    300 µl
  • Seq:
    23
  • Tip style:
    Type L
  • Tip colour:
    Transparent
  • For use with:
    Eppendorf Reference, Research/Research Plus, mLine Mechanical, Proline Mechanical, eLine Electronic
Accessories

Product Information

  • Catalog #
    Colour
    Availability
    Price
  • Tip system box for pipette tip trays, with hinged lid, empty
    Tip system box for pipette tip trays, with hinged lid, empty
    SARS95.1298.001
    Transparent
    Log in for availability
    1 * 1 ST