Order Entry
Belgium
Orders LinkContactUsLinkComponent
Pipette tips

Pipette tips

Type C, 250 µl, +, Transparent, Eppendorf Reference, Research/Research Plus, mLine Mechanical, Proline Mechanical, 96/box, 480/inner case, 1920/case
Catalog # SARS70.760.201
Marketsource
Supplier:   SARSTEDT
Pipette tips
Catalog # SARS70.760.201
Supplier:  SARSTEDT
Marketsource
Pipette tips

Type C, 250 µl, +, Transparent, Eppendorf Reference, Research/Research Plus, mLine Mechanical, Proline Mechanical, 96/box, 480/inner case, 1920/case

Price
Inquire For Price
Log in for availability
1 * 1.920 ST
Product Details & Documents

These high quality tips are suitable for the most common pipettes.

  • Sterile tips are DNase/RNase- and ATP-free, and non pyrogenic
  • Available loose in bags, packed in racks or in Tip StackPack reload systems

Tip StackPack: Tips are preloaded in reusable, stacked 96-tip trays. The trays are designed to safely cover each individual 96-tip layer, shielding the preloaded tips in the stack like a protective screen.

Specifications
  • Packed:
    96/box, 480/inner case, 1920/case
  • Sterile:
    +
  • Volume:
    250 µl
  • Seq:
    08
  • Tip style:
    Type C
  • Tip colour:
    Transparent
  • For use with:
    Eppendorf Reference, Research/Research Plus, mLine Mechanical, Proline Mechanical
Accessories

Product Information

  • Catalog #
    Colour
    Availability
    Price
  • Tip system box for pipette tip trays, with hinged lid, empty
    Tip system box for pipette tip trays, with hinged lid, empty
    SARS95.1298.001
    Transparent
    Log in for availability
    1 * 1 ST

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.