Pipette tips
Type E, 10 µl, +, Transparent, Eppendorf Reference, Research, mLine Mechanical, Proline Mechanical, eLine Electronic, 96/box, 480/inner case, 1920/case
Catalog # SARS70.3010.205
Marketsource
Supplier: SARSTEDT
Jump to:
Product Details & Documents
These high quality tips are suitable for the most common pipettes.
- Sterile tips are DNase/RNase- and ATP-free, and non pyrogenic
- Available loose in bags, packed in racks or in Tip StackPack reload systems
Tip StackPack: Tips are preloaded in reusable, stacked 96-tip trays. The trays are designed to safely cover each individual 96-tip layer, shielding the preloaded tips in the stack like a protective screen.
Specifications
- Packed:96/box, 480/inner case, 1920/case
- Sterile:+
- Volume:10 µl
- Seq:14
- Tip style:Type E
- Tip colour:Transparent
- For use with:Eppendorf Reference, Research, mLine Mechanical, Proline Mechanical, eLine Electronic
Accessories
Product Information
- Catalog #ColourAvailabilityPrice
- Tip system box for pipette tip trays, with hinged lid, emptyTip system box for pipette tip trays, with hinged lid, emptySARS95.1298.001TransparentLog in for availability1 * 1 ST