Order Entry
Belgium
Orders LinkContactUsLinkComponent
Pipette tips

Pipette tips

Type D, 10 µl, -, Transparent, Eppendorf Reference, Research and Research Plus, 1000/bag, 10000/case
Catalog # SARS70.1115
Marketsource
Supplier:   SARSTEDT
Pipette tips
Catalog # SARS70.1115
Supplier:  SARSTEDT
Marketsource
Pipette tips

Type D, 10 µl, -, Transparent, Eppendorf Reference, Research and Research Plus, 1000/bag, 10000/case

Price
Inquire For Price
Log in for availability
1 * 10.000 ST
Product Details & Documents

These high quality tips are suitable for the most common pipettes.

  • Sterile tips are DNase/RNase- and ATP-free, and non pyrogenic
  • Available loose in bags, packed in racks or in Tip StackPack reload systems

Tip StackPack: Tips are preloaded in reusable, stacked 96-tip trays. The trays are designed to safely cover each individual 96-tip layer, shielding the preloaded tips in the stack like a protective screen.

Specifications
  • Packed:
    1000/bag, 10000/case
  • Sterile:
    -
  • Volume:
    10 µl
  • Seq:
    12
  • Tip style:
    Type D
  • Tip colour:
    Transparent
  • For use with:
    Eppendorf Reference, Research and Research Plus
Accessories

Product Information

  • Catalog #
    Colour
    Availability
    Price
  • Tip system box for pipette tip trays, with hinged lid, empty
    Tip system box for pipette tip trays, with hinged lid, empty
    SARS95.1298.001
    Transparent
    Log in for availability
    1 * 1 ST

Aanbevelingen

Aanbevelingen worden alleen gepersonaliseerd op basis van uw winkelvoorkeuren als u daarvoor toestemming hebt gegeven door Verbeter mijn winkelervaring" op Mijn Account pagina" aan te zetten.

Anders ontvangt u algemene aanbevelingen.