Mailing boxes
Mailing boxes with integral restriants for 1 - 4 containers, 198×107×38 mm
Catalog # SARS95.900
Marketsource
Supplier: SARSTEDT
Jump to:
Product Details & Documents
Mailing boxes, made of cardboard, for safe transport of diagnostic specimen in containers.
- With printed label
- Lightweight
- Sturdy
Post-transport packaging for diagnostic samples in accordance with packaging regulation P 650, oversize letter, structural authorisation, flat design.
Specifications
- Description:Mailing boxes with integral restriants for 1 - 4 containers
- Seq:01
- W×D×H:198×107×38 mm