Order Entry
Belgium
Orders LinkContactUsLinkComponent
 

Decontamination Solution, DNA/RNA-ExitusPlus™

DNA/RNA-ExitusPlus™, spray bottle, 500 mL
Catalog # A7089.0500
Supplier:   PanReac AppliChem
Decontamination Solution, DNA/RNA-ExitusPlus™
Catalog # A7089.0500
Supplier:  PanReac AppliChem
CAS Number:  
Decontamination Solution, DNA/RNA-ExitusPlus™
Decontamination Solution, DNA/RNA-ExitusPlus™

DNA/RNA-ExitusPlus™, spray bottle, 500 mL

CAS Number:  
Price
84,30 €
Log in for availability
1 * 500 mL
Product Details & Documents

Solution for the removal of DNA and RNA contamination from surfaces. Suitable for cleaning PCR work stations and equipment, electrophoresis equipment, as well as pipettes and reaction tubes, for example.

  • All components are readily bio-degradable and not harmful or toxic for humans
  • Doesn't contain aggressive mineral acids or alkaline substances
  • No organic solvents or volatile components
  • Delivered as spray bottles or refill bottles (RF)
Specifications

Specifications

  • Pk:
    500 mL
  • Description:
    DNA/RNA-ExitusPlus™, spray bottle
  • Cat. No.:
    A7089.0500
  • Storage temperature:
    Ambient

Specifications