Orders LinkContactUsLinkComponent
Basic cleanroom benches
Basic cleanroom benches
Basic cleanroom bench, 1400×350×450 mm
N° de catalogue 134-0500
Fournisseur:   Palbam
Basic cleanroom benches
N° de catalogue 134-0500
Fournisseur:  Palbam
Basic cleanroom benches
Basic cleanroom benches
Basic cleanroom bench, 1400×350×450 mm
Prix
1 840,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
Cleanroom benches in stainless steel 304 with electropolished finish.
  • Rounded edges
  • Height adjustable legs
  • Leg covers (covers unsightly bolt connections to floor)
  • No snag points
  • Ultra-clean
Delivery information: Typically available from stock for direct shipment from the supplier.
Specifications
  • Description:
    Basic cleanroom bench
  • Seq:
    05
  • W×D×H:
    1400×350×450 mm