Basic cleanroom benches
Basic cleanroom bench, 800×350×450 mm
N° de catalogue 134-0497
Fournisseur: Palbam
Jump to:
Product Details & Documents
Cleanroom benches in stainless steel 304 with electropolished finish.
- Rounded edges
- Height adjustable legs
- Leg covers (covers unsightly bolt connections to floor)
- No snag points
- Ultra-clean
Livraison: Typically available from stock for direct shipment from the supplier.
Documents
Palbam Class - Catalog 2018
Product Literature
Specifications
- Description:Basic cleanroom bench
- Seq:02
- W×D×H:800×350×450 mm