Microplates, opaque OptiPlates™
OptiiPlate™-96 F microplates, 96, Black, Case of 50
Catalog # PERK6055270
Marketsource
Supplier: PerkinElmer
Jump to:
Product Details & Documents
These plates are used for general isotopic, fluorescence, LANCE™, [FP]², AlphaScreen™ or luminescence assays.
- Made of polystyrene
Specifications
- No. of wells:96
- Packed:Case of 50
- Colour:Black
- Description:OptiiPlate™-96 F microplates
- Seq:01