Order Entry
Belgium
Orders LinkContactUsLinkComponent
Baumé hydrometers

Baumé hydrometers

20 - 30, 0,1, 300 mm
Catalog # ALLA0100FG030/15QP
Supplier:   Alla France
Baumé hydrometers
Catalog # ALLA0100FG030/15QP
Supplier:  Alla France
Baumé hydrometers
Baumé hydrometers

20 - 30, 0,1, 300 mm

Price
28,40 €
Log in for availability
1 * 1 ST
Product Details & Documents

Baumé hydrometers calibrated at 15 °C.

Specifications
  • Length:
    300 mm
  • Seq:
    10
  • Division:
    0,1
  • Range:
    20 - 30