Orders LinkContactUsLinkComponent
System Boxes
System Boxes
Transparent, 160×100×75 mm
N° de catalogue 216-3494
System Boxes
N° de catalogue 216-3494
System Boxes
System Boxes
Transparent, 160×100×75 mm
Prix
12,60 €
Log in for availability
1 * 1 ST
Product Details & Documents
Boxes made of transparent glass-clear polystyrene. Suitable for variable applications.
  • Provides immediate overview about content and quantity
  • Large sight opening
  • High stackability
  • Wide labelling pad
  • Excellent stability
Specifications
  • Colour:
    Transparent
  • Dimensions:
    160×100×75 mm
  • Seq:
    01