Orders LinkContactUsLinkComponent
System Boxes
System Boxes
Transparent, 230×150×130 mm
Catalog # 216-3496
System Boxes
Catalog # 216-3496
System Boxes
System Boxes
Transparent, 230×150×130 mm
Price
21,10 €
Log in for availability
1 * 1 ST
Product Details & Documents
Boxes made of transparent glass-clear polystyrene. Suitable for variable applications.
  • Provides immediate overview about content and quantity
  • Large sight opening
  • High stackability
  • Wide labelling pad
  • Excellent stability
Specifications
  • Colour:
    Transparent
  • Dimensions:
    230×150×130 mm
  • Seq:
    02