Order Entry
Belgium
Orders LinkContactUsLinkComponent
Pipette Boxes, Square

Pipette Boxes, Square

65×65×320 mm
Catalog # 612-2043
Pipette Boxes, Square
Catalog # 612-2043
Pipette Boxes, Square
Pipette Boxes, Square

65×65×320 mm

Price
81,90 €
Log in for availability
1 * 1 ST
Product Details & Documents

Anodized aluminium boxes with silicone insert for the protective storage of pipettes.

  • With slip-on cover
  • Autoclavable up to 180 °C
Specifications
  • Seq:
    04
  • W×D×H:
    65×65×320 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.