Order Entry
Belgium
Orders LinkContactUsLinkComponent
Pipette Boxes, Square

Pipette Boxes, Square

65×65×390 mm
Catalog # 612-2044
Pipette Boxes, Square
Catalog # 612-2044
Pipette Boxes, Square
Pipette Boxes, Square

65×65×390 mm

Price
133,00 €
Log in for availability
1 * 1 ST
Product Details & Documents

Anodized aluminium boxes with silicone insert for the protective storage of pipettes.

  • With slip-on cover
  • Autoclavable up to 180 °C
Specifications
  • Seq:
    05
  • W×D×H:
    65×65×390 mm
  • Cat. No.:
    612-2044
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.