Order Entry
Belgium
Orders LinkContactUsLinkComponent
Dissolved oxygen probes, InPro® 6800

Dissolved oxygen probes, InPro® 6800

DO probe, InPro® 6800, InPro® 6800, 0 - 6 μg/l [ppb], Stainless steel, VP, straight, 12×320 mm, Spring water, fruit juices, yeast propagation
Catalog # METN52200967
Supplier:   Mettler - Toledo
Dissolved oxygen probes, InPro® 6800
Catalog # METN52200967
Supplier:  Mettler - Toledo
Dissolved oxygen probes, InPro® 6800

DO probe, InPro® 6800, InPro® 6800, 0 - 6 μg/l [ppb], Stainless steel, VP, straight, 12×320 mm, Spring water, fruit juices, yeast propagation

Price
Inquire For Price
Log in for availability
1 * 1 ST
This product has been blocked by your organization. Please contact your purchasing department for more information
Product Details & Documents

The InPro O2 sensor family provides the means for reliable inline measurement of dissolved oxygen in a wide spectrum of processes that operate under sterile/hygienic conditions.

  • Hygienic design, capable of withstanding CIP procedures
  • Stainless steel surfaces with highly polished finish
  • Steam sterilisable and autoclavable up to 140°C
  • Low detection limit at high signal stability
  • O-rings with FDA and USP Class VI approval
  • Minimal maintenance requirement
  • ISM (Intelligent Sensor Management)
Specifications
  • Concentration:
    0 - 6 μg/l [ppb]
  • Connection:
    VP, straight
  • Ø×L:
    12×320 mm
  • For:
    Spring water, fruit juices, yeast propagation
  • Type:
    InPro® 6800
  • Description:
    DO probe, InPro® 6800
  • Material:
    Stainless steel
  • Seq:
    02
Accessories

Product Information

  • Catalog #
    Availability
    Price
  • Membrane body, T-96, 16 pieces/pack
    Membrane body, T-96, 16 pieces/pack
    METN52206114
    Log in for availability
    1 * 16 ST - 2 510,00 €

Product Information

  • Catalog #
    Availability
    Price
  • Membrane kit, T-96 (4 membrane bodies, 1 O-ring set, 25 ml electrolyte), for InPro 6800/6050
    Membrane kit, T-96 (4 membrane bodies, 1 O-ring set, 25 ml electrolyte), for InPro 6800/6050
    METN52200024
    Log in for availability
    1 * 1 ST - 704,00 €

Recommandations

Les recommandations seront personnalisées en fonction de vos préférences d’achat uniquement si vous avez donné votre consentement en activant l’option " Améliorer mon expérience d’achat " sur la page "Mon compte".

Dans le cas contraire, vous recevrez des recommandations génériques.