Order Entry
Belgium
Orders LinkContactUsLinkComponent
 

Water

Plastic bottle, 1 L
Catalog # KRAF10791.3000
Supplier:   Bernd Kraft
Water
Catalog # KRAF10791.3000
Supplier:  Bernd Kraft
CAS Number:  7732-18-5
Water

Plastic bottle, 1 L

CAS Number:  7732-18-5
Price
63,10 €
Log in for availability
1 * 1 L
Product Details & Documents
Synonyms:  Hydrogen oxide
  • CAS Number:7732-18-5
  • Formula:H₂O
  • MDL Number:MFCD00011332
  • Molecular Weight:18.02 g/mol
  • Merck Index:13,10098
  • Boiling Pt:100 °C (1013 hPa)
  • Melting Pt:0 °C
  • Density:1 g/cm³ (4 °C)
  • Storage Temperature:Ambient
  • EINECS:231-791-2
Specifications

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    1 L

Specifications