Order Entry
Belgium
Orders LinkContactUsLinkComponent
Buffer, TISAB solution (Total Ionic Strength Adjustment Buffer) for fluoride determinations

Buffer, TISAB solution (Total Ionic Strength Adjustment Buffer) for fluoride determinations

TISAB solution (Total Ionic Strength Adjustment Buffer) for fluoride determinations, Plastic bottle, 1 L
Catalog # KRAF10336.3000
Supplier:   Bernd Kraft
Buffer, TISAB solution (Total Ionic Strength Adjustment Buffer) for fluoride determinations
Catalog # KRAF10336.3000
Supplier:  Bernd Kraft
CAS Number:  
Buffer, TISAB solution (Total Ionic Strength Adjustment Buffer) for fluoride determinations

TISAB solution (Total Ionic Strength Adjustment Buffer) for fluoride determinations, Plastic bottle, 1 L

CAS Number:  
Price
Inquire For Price
Log in for availability
1 * 1 L
Product Details & Documents
Specifications

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    1 L
  • Description:
    TISAB solution (Total Ionic Strength Adjustment Buffer) for fluoride determinations

Specifications