Order Entry
Belgium
Orders LinkContactUsLinkComponent
 

Buffer solution pH 10

Buffer solution, 10, Ammonia, 20 L
Catalog # KRAF03596.7020
Supplier:   Bernd Kraft
Buffer solution pH 10
Catalog # KRAF03596.7020
Supplier:  Bernd Kraft
CAS Number:  
Buffer solution pH 10

Buffer solution, 10, Ammonia, 20 L

CAS Number:  
Price
Inquire For Price
Log in for availability
1 * 20 L
Product Details & Documents

This solution is a combination of 53,5 g NH₄Cl, 350 ml NH₃ solution 25% and Cu-EDTA/l used for complexometric determination of zinc.

Specifications

Specifications

  • pH buffer type:
    Ammonia
  • Pk:
    20 L
  • Description:
    Buffer solution
  • pH value:
    10

Specifications