Order Entry
Belgium
Orders LinkContactUsLinkComponent
 

Buffer solutions, ready to use

13,0, Glycine, Colourless, 20 °C, Plastic bottle, 1 L
Catalog # KRAF03096.3060
Supplier:   Bernd Kraft
Buffer solutions, ready to use
Catalog # KRAF03096.3060
Supplier:  Bernd Kraft
CAS Number:  
Buffer solutions, ready to use
Buffer solutions, ready to use

13,0, Glycine, Colourless, 20 °C, Plastic bottle, 1 L

CAS Number:  
Price
41,30 €
Log in for availability
1 * 1 L
Product Details & Documents
  • NIST traceable

Documents

Specifications

Specifications

  • Pack type:
    Plastic bottle
  • pH buffer type:
    Glycine
  • Pk:
    1 L
  • Reference temperature:
    20 °C
  • Colour:
    Colourless
  • pH value:
    13,0

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.