Order Entry
Belgium
Orders LinkContactUsLinkComponent
 

Sample cuvettes

Sample cuvettes, with lid, matched pair, 3,5 ml, 10 mm, Glass
Catalog # 705-0084
Supplier:   Hach
Sample cuvettes
Catalog # 705-0084
Supplier:  Hach
Sample cuvettes
Sample cuvettes

Sample cuvettes, with lid, matched pair, 3,5 ml, 10 mm, Glass

Price
320,00 €
Log in for availability
1 * 2 ST
Product Details & Documents

Sample cuvettes for photometric measurements.

  • Available with or without lid
  • Available reusable (made from glass) or for single use (made from plastics)
Specifications
  • Path length:
    10 mm
  • Volume:
    3,5 ml
  • Window material:
    Glass
  • Description:
    Sample cuvettes, with lid, matched pair
  • Seq:
    04
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.