Labels, hazard warning
Hazard labels: low temperature, PIC 323-TRI 100-B7541, Polyester, Black on yellow, 100×87 mm, 3 labels/card
Catalog # 116-0187
Supplier: Brady
Jump to:
Product Details & Documents
These self adhesive signs are designed to warn people to be careful and to take precautions of potential hazards.
Available in different sizes.
: Conforms to Health and Safety (Safety Signs and Signals) Regulations 1996.
Specifications
- Colour:Black on yellow
- Description:Hazard labels: low temperature
- Dimensions:100×87 mm
- Material:Polyester
- Model:PIC 323-TRI 100-B7541
- Packed:3 labels/card