Orders LinkContactUsLinkComponent
Nalgene™ Screw Caps
Nalgene™ Screw Caps
Linerless, 20-415, Natural, PP
Catalog # 215-1597
   Low Manufacturing Impact
Nalgene™ Screw Caps
Catalog # 215-1597
   Low Manufacturing Impact
Nalgene™ Screw Caps
Nalgene™ Screw Caps
Linerless, 20-415, Natural, PP
Price
725,00 €
Log in for availability
1 * 2.000 ST
Product Details & Documents
PP, coloured.
  • Very good chemical resistance
  • Heat resistant up to 135 °C
  • Autoclavable
Screw caps without inserts for Nalgene® narrow and wide mouth bottles, as well as Erlenmeyer flasks with thread.
Specifications
  • Cap Size:
    20-415
  • Colour:
    Natural
  • Description:
    Linerless
  • Material:
    PP
  • Seq:
    02
  • Cat. No.:
    215-1597
  • Environmental certification:
    NAR Certificate of Retirement
  • Environmentally Preferable:
    Low Manufacturing Impact