Labels, biohazard
Biohazard labels, PIC 312-Tri 100-B7527, PP, Black on yellow, 100×87 mm, 1 label/card
Catalog # 116-0171
Supplier: Brady
Jump to:
Product Details & Documents
All signs comply with current European legislation.
Ordering information: The polyester labels have a permanent acrylic adhesive.
Specifications
- Colour:Black on yellow
- Description:Biohazard labels
- Dimensions:100×87 mm
- Material:PP
- Seq:1
- Model:PIC 312-Tri 100-B7527
- Packed:1 label/card
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



