Order Entry
Belgium
Orders LinkContactUsLinkComponent
Sample containers with screw cap

Sample containers with screw cap

30 ml, Sterile, with white spoon and label, White, 25 mm, 90 mm, Individually wrapped
Catalog # KART5623
Supplier:   KARTELL
Sample containers with screw cap
Catalog # KART5623
Supplier:  KARTELL
Sample containers with screw cap
Sample containers with screw cap

30 ml, Sterile, with white spoon and label, White, 25 mm, 90 mm, Individually wrapped

Price
371,00 €
Log in for availability
1 * 400 ST
Product Details & Documents

Made of PS, with PE screw cap and spoon.

  • Disposable
  • Available sterile or non sterile
  • Good chemical resistance
  • Lightweight
Specifications
  • Ext.Ø:
    25 mm
  • Packed:
    Individually wrapped
  • Type:
    Sterile, with white spoon and label
  • Capacity:
    30 ml
  • Height:
    90 mm
  • Seq:
    01
  • Cap colour:
    White
Recommandations

Les recommandations seront personnalisées en fonction de vos préférences d’achat uniquement si vous avez donné votre consentement en activant l’option " Améliorer mon expérience d’achat " sur la page "Mon compte".

Dans le cas contraire, vous recevrez des recommandations génériques.