Orders LinkContactUsLinkComponent
Transport containers
Transport containers
Slide mailers, PP, 87×47×16 mm, 2
Catalog # DELT900025
Supplier:   DELTALAB
Transport containers
Catalog # DELT900025
Supplier:  DELTALAB
Transport containers
Transport containers
Slide mailers, PP, 87×47×16 mm, 2
Price
56,40 €
Log in for availability
1 * 100 ST
Product Details & Documents
Designed for slide transport and protection.
  • Made of polypropylene
  • Easy snap-lock
  • Not indicated for liquid samples
Compartment shape ensures tight fit: no risk of damaging slides and to send by mail, recommend to use special envelopes.
Documents
Specifications
  • No. of slides:
    2
  • Description:
    Slide mailers
  • Material:
    PP
  • Seq:
    02
  • W×D×H:
    87×47×16 mm