Pipette tips
Gilson type, bevelled, 2 - 200 µl, -, Yellow, Gilson P20, P50, P100, P200, Bulk (25×1000)
Supplier: DELTALAB
Jump to:
Product Details & Documents
Documents
Specifications
- Packed:Bulk (25×1000)
- Sterile:-
- Volume:2 - 200 µl
- Seq:07
- Tip style:Gilson type, bevelled
- Tip colour:Yellow
- For use with:Gilson P20, P50, P100, P200