Orders LinkContactUsLinkComponent
Cavity slides
Cavity slides
Cavity slides, 3
  630-1613
 :   MARIENFELD
Cavity slides
  630-1613
Cavity slides
Cavity slides
Cavity slides, 3
231,00 €
1 * 50 ST
Soda-lime glass of hydrolytic class 3 cavity slides.
Carefully ground and polished for in vitro diagnostic (IVD) applications.
Certifications: IVD 98/79/EC
Documents
  • No. of wells:
    3
  • Description:
    Cavity slides
  • Seq:
    04