Rods, AR-Glas®
5±0,15 mm, 1500 mm
Catalog # SCOR1028415
Supplier: Schott AG
Jump to:
Product Details & Documents
Made of AR®-Glas.
- Soft glass is acid-resistant
- Has high thermal expansion for pressure melting and for fusing with platinum or dumet and other alloys
Specifications
- Ext.Ø:5±0,15 mm
- Length:1500 mm
- Seq:03
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



