Order Entry
Belgium
Orders LinkContactUsLinkComponent
Scoops

Scoops

375 ml, 290 mm
Catalog # USBE3723
Supplier:   USBECK Laborgeräte
Scoops
Catalog # USBE3723
Scoops
Scoops

375 ml, 290 mm

Price
11,30 €
Log in for availability
1 * 1 ST
Product Details & Documents

Aluminium, smooth. For collecting or measuring powders, granulates or pastes.

  • Good chemical resistance
  • Excellent thermal resistance
  • Light, robust and easy to use
Specifications
  • Capacity:
    375 ml
  • Length:
    290 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.