Orders LinkContactUsLinkComponent
Particle traps for use with PLOT columns
Particle traps for use with PLOT columns
Particle trap for PoraPLOT columns, fused silica, Int.Ø×L: 0,53 mm × 2,5 m
Catalog # VARICP4017
Supplier:   VARIAN
Particle traps for use with PLOT columns
Catalog # VARICP4017
Supplier:  VARIAN
Particle traps for use with PLOT columns
Particle trap for PoraPLOT columns, fused silica, Int.Ø×L: 0,53 mm × 2,5 m
Price
193,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
Though highly stabilised, it is impossible to guarantee that no particles will dislodge from the column wall. When used in valve-switching applications, the use of a particle trap can prevent scarring of the rotors.
Specifications
  • Description:
    Particle trap for PoraPLOT columns, fused silica, Int.Ø×L: 0,53 mm × 2,5 m
Recommandations
Les recommandations seront personnalisées en fonction de vos préférences d’achat uniquement si vous avez donné votre consentement en activant l’option " Améliorer mon expérience d’achat " sur la page "Mon compte".
Dans le cas contraire, vous recevrez des recommandations génériques.