Saisie de commande
Belgium
Lien pour commandesContactUsLinkComponent
Solid phase microextraction fibers
Solid phase microextraction fibers
SPME fiber for autosampler use, polyacrylate, 24 gauge, 85 µm fiber coating
Catalog # VARI391896306
Supplier:   VARIAN
Solid phase microextraction fibers
Catalog # VARI391896306
Supplier:  VARIAN
Solid phase microextraction fibers
SPME fiber for autosampler use, polyacrylate, 24 gauge, 85 µm fiber coating
Price
Inquire For Price
Log in for availability
1 * 3 ST
Product Details & Documents
Solid phase microextraction (SPME) is a technique for extracting analytes from solid, liquid or gaseous samples by adsorbing them onto the SPME fiber and then desorbing them into an inlet, either on a gas chromatograph (GC) or an HPLC system. SPME is amenable to automation using an autosampler or it can be performed manually as well.
Solid phase microextraction kits: Kits are also available to support method development, offering a variety of fiber types and configurations within a single kit. SPME fiber kits contain three fibers.
Specifications
  • Désignation:
    SPME fiber for autosampler use, polyacrylate, 24 gauge, 85 µm fiber coating
  • Seq:
    05
Accessories
Product Information
  • Catalog #
    Désignation
    Availability
    Price
  • VARI392609902
    Merlin microseal, SPME replacement seal, 23 gauge
    Log in for availability
    1 * 1 ST - 416,00 €
Product Information
  • Catalog #
    Désignation
    Availability
    Price
  • VARISU57347U
    SPME fiber holder for CTC autosampler
    Log in for availability
    1 * 1 ST
Product Information
  • Catalog #
    Désignation
    Availability
    Price
  • VARISU57357U
    SPME stand, 15 ml
    Log in for availability
    1 * 1 ST
Product Information
  • Catalog #
    Désignation
    Availability
    Price
  • VARISU57356U
    SPME inlet guide for manual injection
    Log in for availability
    1 * 1 ST