Order Entry
Belgium
Orders LinkContactUsLinkComponent
Solid phase microextraction fibers

Solid phase microextraction fibers

SPME fiber for manual use, carboxen/PDMS, 23 gauge, 75 µm fiber coating
Catalog # VARISU57344U
Supplier:   VARIAN
Solid phase microextraction fibers
Catalog # VARISU57344U
Supplier:  VARIAN
Solid phase microextraction fibers

SPME fiber for manual use, carboxen/PDMS, 23 gauge, 75 µm fiber coating

Price
Inquire For Price
Log in for availability
1 * 3 ST
Product Details & Documents

Solid phase microextraction (SPME) is a technique for extracting analytes from solid, liquid or gaseous samples by adsorbing them onto the SPME fiber and then desorbing them into an inlet, either on a gas chromatograph (GC) or an HPLC system. SPME is amenable to automation using an autosampler or it can be performed manually as well.

Solid phase microextraction kits: Kits are also available to support method development, offering a variety of fiber types and configurations within a single kit. SPME fiber kits contain three fibers.

Specifications
  • Description:
    SPME fiber for manual use, carboxen/PDMS, 23 gauge, 75 µm fiber coating
  • Seq:
    20
Accessories

Product Information

  • Catalog #
    Availability
    Price
  • Merlin microseal, SPME replacement seal, 23 gauge
    Merlin microseal, SPME replacement seal, 23 gauge
    VARI392609902
    Log in for availability
    1 * 1 ST - 416,00 €

Product Information

  • Catalog #
    Availability
    Price
  • SPME fiber holder for CTC autosampler
    SPME fiber holder for CTC autosampler
    VARISU57347U
    Log in for availability
    1 * 1 ST

Product Information

  • Catalog #
    Availability
    Price
  • SPME stand, 15 ml
    SPME stand, 15 ml
    VARISU57357U
    Log in for availability
    1 * 1 ST

Product Information

  • Catalog #
    Availability
    Price
  • SPME inlet guide for manual injection
    SPME inlet guide for manual injection
    VARISU57356U
    Log in for availability
    1 * 1 ST