Order Entry
Belgium
Orders LinkContactUsLinkComponent
Solid phase microextraction fibers

Solid phase microextraction fibers

SPME fiber for autosampler use, carboxen/PDMS, 85 µm fiber coating, Stableflex
Catalog # VARISU57335U
Supplier:   VARIAN
Solid phase microextraction fibers
Catalog # VARISU57335U
Supplier:  VARIAN
Solid phase microextraction fibers

SPME fiber for autosampler use, carboxen/PDMS, 85 µm fiber coating, Stableflex

Price
Inquire For Price
Log in for availability
1 * 3 ST
Product Details & Documents

Solid phase microextraction (SPME) is a technique for extracting analytes from solid, liquid or gaseous samples by adsorbing them onto the SPME fiber and then desorbing them into an inlet, either on a gas chromatograph (GC) or an HPLC system. SPME is amenable to automation using an autosampler or it can be performed manually as well.

Solid phase microextraction kits: Kits are also available to support method development, offering a variety of fiber types and configurations within a single kit. SPME fiber kits contain three fibers.

Specifications
  • Description:
    SPME fiber for autosampler use, carboxen/PDMS, 85 µm fiber coating, Stableflex
  • Seq:
    06
Accessories

Product Information

  • Catalog #
    Availability
    Price
  • Merlin microseal, SPME replacement seal, 23 gauge
    Merlin microseal, SPME replacement seal, 23 gauge
    VARI392609902
    Log in for availability
    1 * 1 ST - 416,00 €

Product Information

  • Catalog #
    Availability
    Price
  • SPME fiber holder for CTC autosampler
    SPME fiber holder for CTC autosampler
    VARISU57347U
    Log in for availability
    1 * 1 ST

Product Information

  • Catalog #
    Availability
    Price
  • SPME stand, 15 ml
    SPME stand, 15 ml
    VARISU57357U
    Log in for availability
    1 * 1 ST

Product Information

  • Catalog #
    Availability
    Price
  • SPME inlet guide for manual injection
    SPME inlet guide for manual injection
    VARISU57356U
    Log in for availability
    1 * 1 ST

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.