Order Entry
Belgium
Orders LinkContactUsLinkComponent
Solid phase extraction cartridges, Bond Elut™ PBA

Solid phase extraction cartridges, Bond Elut™ PBA

Straight barrel cartridge, PBA, 60 Å, 40 µm, 3 ml, 200/60 mg
Catalog # VARI12105033
Supplier:   VARIAN
Solid phase extraction cartridges, Bond Elut™ PBA
Catalog # VARI12105033
Supplier:  VARIAN
Solid phase extraction cartridges, Bond Elut™ PBA

Straight barrel cartridge, PBA, 60 Å, 40 µm, 3 ml, 200/60 mg

Price
295,00 €
Log in for availability
1 * 50 ST
Product Details & Documents

PBA is a unique sorbent consisting of a phenylboronic acid covalently linked to a silica gel surface. Covalent processes result in a higher retention to the sorbent surface than other mechanisms, resulting in superior clean-up. The boronate group has a high specificity for cis-diol containing compounds such as catechols, nucleic acids, low molecular weight proteins and carbohydrates.

  • High specificity for cis-diol compounds
  • Very effective in separating DNA and RNA

Bond Elut™ PBA can also be used to retain oligomeric materials such as polyethylene glycols (PEGs) from a variety of aqueous or biological matrices.

Specifications
  • Particle Size:
    40 µm
  • Pore size:
    60 Å
  • Capacity:
    3 ml
  • Description:
    Straight barrel cartridge
  • Seq:
    06
  • Bed weight:
    200/60 mg
  • Phase:
    PBA
  • Carbon load:
    7,9%
  • Cat. No.:
    VARI12105033
  • Endcapped:
    No
  • Surface area:
    500 m²/g