Saisie de commande
Belgium
Lien pour commandesContactUsLinkComponent
Solid phase extraction cartridges, Bond Elut™ PH

Solid phase extraction cartridges, Bond Elut™ PH

Cartouches de LRC, PH, 60 Å, 120 µm, 10 ml, 100 mg
Catalog # VARI14113005
Supplier:   VARIAN
Solid phase extraction cartridges, Bond Elut™ PH
Catalog # VARI14113005
Supplier:  VARIAN
Solid phase extraction cartridges, Bond Elut™ PH

Cartouches de LRC, PH, 60 Å, 120 µm, 10 ml, 100 mg

Price
146,00 €
Log in for availability
1 * 50 ST
Product Details & Documents

Bond Elut™ PH is a non-polar bonded silica material which exhibits a different selectivity to alkyl or aliphatic functionalised phases such as C8 or cyclohexyl. The electron density present in the aromatic ring affords an enhancement in the retention of conjugated or aromatic ring-containing analytes due to desirable pi-pi interactions.

  • Added selectivity compared to other non-polar sorbents
  • Enhanced retention of planar, conjugated organic molecules
  • Similar polarity to C8
Specifications
  • Granulométrie:
    120 µm
  • Taille de pore:
    60 Å
  • Capacité:
    10 ml
  • Désignation:
    Cartouches de LRC
  • Seq:
    02
  • Poids du lit:
    100 mg
  • Greffage:
    PH
  • Cat. No.:
    VARI14113005
  • End-capping:
    Oui
  • Surface area:
    500 m²/g
  • Charge de carbone:
    10,7%
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.