Order Entry
Belgium
Orders LinkContactUsLinkComponent
Solid phase extraction cartridges, Bond Elut™ PH

Solid phase extraction cartridges, Bond Elut™ PH

Straight barrel cartridge, PH, 60 Å, 120 µm, 1 ml, 100 mg
Catalog # VARI14102005
Supplier:   VARIAN
Solid phase extraction cartridges, Bond Elut™ PH
Catalog # VARI14102005
Supplier:  VARIAN
Solid phase extraction cartridges, Bond Elut™ PH

Straight barrel cartridge, PH, 60 Å, 120 µm, 1 ml, 100 mg

Price
235,00 €
Connexion Pour la disponibilité
1 * 100 ST
Product Details & Documents

Bond Elut™ PH is a non-polar bonded silica material which exhibits a different selectivity to alkyl or aliphatic functionalised phases such as C8 or cyclohexyl. The electron density present in the aromatic ring affords an enhancement in the retention of conjugated or aromatic ring-containing analytes due to desirable pi-pi interactions.

  • Added selectivity compared to other non-polar sorbents
  • Enhanced retention of planar, conjugated organic molecules
  • Similar polarity to C8
Spécifications
  • Particle Size:
    120 µm
  • Pore size:
    60 Å
  • Capacity:
    1 ml
  • Description:
    Straight barrel cartridge
  • Seq:
    09
  • Bed weight:
    100 mg
  • Phase:
    PH
  • Carbon load:
    10,7%
  • Cat. No.:
    VARI14102005
  • Endcapped:
    Yes
  • Surface area:
    500 m²/g
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.