Orders LinkContactUsLinkComponent
Solid phase extraction cartridges, Bond Elut™ SI
Solid phase extraction cartridges, Bond Elut™ SI
120
Catalog # VARI14113036
Supplier:   VARIAN
Solid phase extraction cartridges, Bond Elut™ SI
Catalog # VARI14113036
Supplier:  VARIAN
Solid phase extraction cartridges, Bond Elut™ SI
120
Price
252,00 €
Log in for availability
1 * 50 ST
Product Details & Documents
Native silica is generally regarded as the most polar SPE sorbent available. Bond Elut™ SI is particularly effective at separating compounds with a very similar structure. Applying the analytes in a non-polar solvent, then increasing the solvent polarity by increasing the concentration of a polar modifier, such as THF or ethyl acetate, delivers effective separations.
  • Highly polar phase retains polar molecules from non-polar matrices
  • High purity silica
  • Separate compounds with very similar structures
Specifications
  • Particle Size:
    120
Recommandations
Les recommandations seront personnalisées en fonction de vos préférences d’achat uniquement si vous avez donné votre consentement en activant l’option " Améliorer mon expérience d’achat " sur la page "Mon compte".
Dans le cas contraire, vous recevrez des recommandations génériques.