Micro pipette tips, OMIX®
OMIX, C4, 2 - 10 µl, 6 racks, 96 each
Catalog # VARIA5700910K
Supplier: VARIAN
Jump to:
Product Details & Documents
Bond Elut OMIX pipette tips reliably purify and enrich femtomole and picomole levels of peptides and proteins prior to MALDI-TOF or LC/MS/MS. The unique monolithic sorbent technology used in Bond Elut OMIX consistently outperforms other tips by delivering uniform flow and strong analyte-to-surface interactions. Bond Elut OMIX tips bind more peptides to deliver better sequence coverage.
- Fast, uniform flow maximizes productivity and reproducibility
- Minimal peptide losses lead to higher recoveries
- Available in three phases and sizes to deliver better sequence coverage
Specifications
- Packed:6 racks, 96 each
- Volume:2 - 10 µl
- Seq:08
- Tip style:OMIX, C4