Orders LinkContactUsLinkComponent
Micro pipette tips, OMIX®
Micro pipette tips, OMIX®
OMIX Mini-Bed, C18, 0,5 - 2 µl, 6 racks, 96 each
Catalog # VARIA57003MBK
Supplier:   VARIAN
Micro pipette tips, OMIX®
Catalog # VARIA57003MBK
Supplier:  VARIAN
Micro pipette tips, OMIX®
Micro pipette tips, OMIX®
OMIX Mini-Bed, C18, 0,5 - 2 µl, 6 racks, 96 each
Price
1 690,00 €
Log in for availability
1 * 576 ST
Product Details & Documents
Bond Elut OMIX pipette tips reliably purify and enrich femtomole and picomole levels of peptides and proteins prior to MALDI-TOF or LC/MS/MS. The unique monolithic sorbent technology used in Bond Elut OMIX consistently outperforms other tips by delivering uniform flow and strong analyte-to-surface interactions. Bond Elut OMIX tips bind more peptides to deliver better sequence coverage.
  • Fast, uniform flow maximizes productivity and reproducibility
  • Minimal peptide losses lead to higher recoveries
  • Available in three phases and sizes to deliver better sequence coverage
Specifications
  • Packed:
    6 racks, 96 each
  • Volume:
    0,5 - 2 µl
  • Seq:
    05
  • Tip style:
    OMIX Mini-Bed, C18