Orders LinkContactUsLinkComponent
Tubes and systems for active sampling
Activated charcoal tubes, G
  DRAE6400746
 :   Dräger
Tubes and systems for active sampling
  DRAE6400746
 :  Dräger
Tubes and systems for active sampling
Tubes and systems for active sampling
Activated charcoal tubes, G
1 100,00 €
1 * 200 ST
Sampling activated tubes for direct measurement indication even with complex compounds and mixtures of substances in air investigations and gas analyses.
  • High precision guaranteed
  • Very reliable in use
  • Improved colour depth and contrast using an optimised production method
  • Type:
    G
  • Description:
    Activated charcoal tubes
  • Seq:
    09