Buffer solutions, colour coded, Fluka™
10,00, Violet, 20 °C, Plastic bottle, 0,5 L, 500 mL
Supplier: Honeywell Chemicals
Buffer solutions, colour coded, Fluka™
Catalog # 33668-500ML
Supplier: Honeywell Chemicals
CAS Number:
Jump to:
Product Details & Documents
These buffer solutions are colour-coded for easy identification and the pH 4 and 7 version have fungicide additive.
Certificates
Search by Lot NumberSpecifications
Specifications
- Pack type:Plastic bottle
- Pk:500 mL
- Reference temperature:20 °C
- Colour:Violet
- Pk Info:0,5 L
- pH value:10,00
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



