Orders LinkContactUsLinkComponent
Urea, Reference Standards
Urea, Reference Standards
Urea, USP reference standard, Vial, 200 mg
Catalog # 1706698.
Supplier:   USP
Urea, Reference Standards
Catalog # 1706698.
Supplier:  USP
CAS Number:  
Urea, Reference Standards
Urea, Reference Standards
Urea, USP reference standard, Vial, 200 mg
CAS Number:  
Price
361,00 €
Log in for availability
1 * 200 mg
Product Details & Documents
USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.
Certifications: US Pharmacopea Reference Standard
Specifications
Specifications
  • Pack type:
    Vial
  • Pk:
    200 mg
  • Standards:
    USP reference standard
  • Description:
    Urea
  • Cat. No.:
    1706698.
  • Description:
    Urea
  • CAS No.:
    57-13-6
Specifications
Recommandations
Les recommandations seront personnalisées en fonction de vos préférences d’achat uniquement si vous avez donné votre consentement en activant l’option " Améliorer mon expérience d’achat " sur la page "Mon compte".
Dans le cas contraire, vous recevrez des recommandations génériques.