Order Entry
Belgium
Orders LinkContactUsLinkComponent
Polyethylene, Reference Standards

Polyethylene, Reference Standards

Low-Density Polyethylene, USP reference standard, 3 strips, Strips, 1 KIT
Catalog # 1546809.
Supplier:   USP
Polyethylene, Reference Standards
Catalog # 1546809.
Supplier:  USP
CAS Number:  
Polyethylene, Reference Standards

Low-Density Polyethylene, USP reference standard, 3 strips, Strips, 1 KIT

CAS Number:  
Price
447,00 €
Log in for availability
1 * 1 KIT
Product Details & Documents

USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.

Certifications: US Pharmacopea Reference Standard

Documents

Specifications

Specifications

  • Pack type:
    Strips
  • Pk:
    1 KIT
  • Standards:
    USP reference standard
  • Description:
    Low-Density Polyethylene
  • Pk Info:
    3 strips
  • Cat. No.:
    1546809.
  • Formula:
    C₂H₄
  • Description:
    Low-Density Polyethylene
  • CAS No.:
    9002-88-4

Specifications