Saisie de commande
Belgium
Lien pour commandesContactUsLinkComponent
Phenoxybenzamine, Reference Standards

Phenoxybenzamine, Reference Standards

Phenoxybenzamine Hydrochloride, USP reference standard, Vial, 250 mg
Catalog # 1526007.
Supplier:   USP
Phenoxybenzamine, Reference Standards
Catalog # 1526007.
Supplier:  USP
CAS Number:  
Phenoxybenzamine, Reference Standards

Phenoxybenzamine Hydrochloride, USP reference standard, Vial, 250 mg

CAS Number:  
Price
375,00 €
Log in for availability
1 * 250 mg
Product Details & Documents

USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.

Certifications: US Pharmacopea Reference Standard

Caution: Cold Shipment required

Specifications

Specifications

  • Type de conditionnement:
    Vial
  • Cdt:
    250 mg
  • Normes:
    USP reference standard
  • Désignation:
    Phenoxybenzamine Hydrochloride
  • UN No.:
    3261
  • Cat. No.:
    1526007.
  • N ° CAS.:
    63-92-3
  • Désignation:
    Phenoxybenzamine Hydrochloride

Specifications