Cover glasses
#1, 18 mm
HECH41001118
Borosilicate glass highly resistant cover glasses with precise and faultless cut.
: Available in different diameters.
- Ø:18 mm
- No.:#1
- Seq:09
Borosilicate glass highly resistant cover glasses with precise and faultless cut.
: Available in different diameters.