Order Entry
Belgium
Orders LinkContactUsLinkComponent
Stirring rods

Stirring rods

Stirring rods, glass, with fused ends, Glass, 4 mm, 200 mm
Catalog # 441-0341
Stirring rods
Catalog # 441-0341
Stirring rods
Stirring rods

Stirring rods, glass, with fused ends, Glass, 4 mm, 200 mm

Price
40,30 €
Log in for availability
1 * 100 ST
Product Details & Documents

AR-Glas®. With fused ends.

Specifications
  • Ø:
    4 mm
  • Description:
    Stirring rods, glass, with fused ends
  • Length:
    200 mm
  • Material:
    Glass
  • Seq:
    015
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.