Order Entry
Belgium
Orders LinkContactUsLinkComponent
Rectangular cover glasses

Rectangular cover glasses

45×60 mm, 0,13 - 0,16 mm
Catalog # HECH92099001280
Rectangular cover glasses
Catalog # HECH92099001280
Rectangular cover glasses
Rectangular cover glasses

45×60 mm, 0,13 - 0,16 mm

Price
307,00 €
Log in for availability
1 * 1.000 ST
Product Details & Documents

Cover glass D 263® M made of borosilicate glass.

  • Free from colour distortion and highly transparent
  • Excellent quality, low levels of inclusions, striae, bubbles and streaks
  • High spectral transmission, excellent flatness, optimum resistance to chemicals

Certifications: ISO 8255

Specifications
  • L×W:
    45×60 mm
  • Seq:
    014
  • Thickness:
    0,13 - 0,16 mm
Recommandations

Les recommandations seront personnalisées en fonction de vos préférences d’achat uniquement si vous avez donné votre consentement en activant l’option " Améliorer mon expérience d’achat " sur la page "Mon compte".

Dans le cas contraire, vous recevrez des recommandations génériques.