Orders LinkContactUsLinkComponent
Inlet liners
PTV liner, high temperature, borosilicate, 668 µl, 3,4 mm
  HEWL5188-5356
 :   Agilent
Inlet liners
  HEWL5188-5356
 :  Agilent
Inlet liners
Inlet liners
PTV liner, high temperature, borosilicate, 668 µl, 3,4 mm
149,00 €
1 * 1 ST
These inlet liners prevent adsorption and ensure accurate sample transfer onto the GC column through a robust deactivated surface. These liners are recommended for trace level analysis of active analytes. Liners with inert wool retain non-volatiles, prolonging column life and lowering frequency of MS source maintenance.
  • Consistent high level of inertness
  • Ø int.:
    3,4 mm
  • Description:
    PTV liner, high temperature, borosilicate, 668 µl
  • Seq:
    63