Orders LinkContactUsLinkComponent
Half area microplates, 96-well
Half area, solid bottom, non treated surface, Clear, -
  738-0145
   Low Manufacturing Impact
Half area microplates, 96-well
  738-0145
   Low Manufacturing Impact
Half area microplates, 96-well
Half area microplates, 96-well
Half area, solid bottom, non treated surface, Clear, -
118,00 €
1 * 40 ST
PS, without lid. 96-well half area microplates offer an alternative to 384-well or 1536-well microplates when low sample volume is an important criterion but automation is not necessarily appropriate. They can be pipetted manually while at the same time allow a reduction in sample volume of up to 50%.
Microplates with clear bottoms feature micro-thin µClear® bottoms for exceptional optical properties and standardised pathlength, eliminating the need for pathlength determinations. White plates are ideal for luminescence assays, while black plates are suitable for fluorescence assays. Both formats are also suitable for cell-based assays.
  • Sterile:
    -
  • Colour:
    Clear
  • Description:
    Half area, solid bottom, non treated surface
  • Seq:
    02
  • Cat. No.:
    738-0145
  • Environmental certification:
    ISO 14001 (241002); ISO 50001 (241002)
  • Environmentally Preferable:
    Low Manufacturing Impact
Recommandations
Les recommandations seront personnalisées en fonction de vos préférences d’achat uniquement si vous avez donné votre consentement en activant l’option " Améliorer mon expérience d’achat " sur la page "Mon compte".
Dans le cas contraire, vous recevrez des recommandations génériques.