Order Entry
Belgium
Orders LinkContactUsLinkComponent
Curcumin

Curcumin

Glass bottle for solids, 10 g
Catalog # CAYM81025-10
Supplier:   Cayman Chemical
Curcumin
Catalog # CAYM81025-10
Supplier:  Cayman Chemical
CAS Number:  458-37-7
Curcumin

Glass bottle for solids, 10 g

CAS Number:  458-37-7
Price
108,00 €
Log in for availability
1 * 10 g
Product Details & Documents
Synonyms:  (1E,6E)-1,7-Bis(4-hydroxy-3-methoxyphenyl)hepta-1,6-diene-3,5-dione
  • CAS Number:458-37-7
  • Formula:C₂₁H₂₀O₆
  • MDL Number:MFCD00008365
  • Molecular Weight:368.39 g/mol
  • EINECS:207-280-5

Documents

Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    10 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.